CAS RN 17957-94-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(+)-Menthofuran, ≥99% (sum of enantiomers, GC), MilliporeSigma™ Supelco™
(+)-Menthofuran is a metabolite of pulegone, found in mint plants, which is basically produced from (+)-pulegone by menthofuran synthase enzyme, under certain conditions of stress like low sunlight, limited moisture and nutrient deficiencies.