Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-Menthofuran, ≥99% (sum of enantiomers, GC), MilliporeSigma™ Supelco™

Catalog No. 111013369 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3369 1 mL
11-101-3368 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-3369 Supplier MilliporeSigma Supelco Supplier No. 636611MLF
Add to Cart
Add to Cart

(+)-Menthofuran is a metabolite of pulegone, found in mint plants, which is basically produced from (+)-pulegone by menthofuran synthase enzyme, under certain conditions of stress like low sunlight, limited moisture and nutrient deficiencies.

Featured Industries: Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

CAS 17957-94-7
Density 0.97 g/mL (at 20°C)
Physical Form Neat
Linear Formula C10H14 O
MDL Number MFCD00142332
Boiling Point 204°C to 206°C
Synonym (6 R)-4,5,6,7-Tetrahydro-3,6-dimethylbenzofuran; (R)-3,6-Dimethyl-4,5,6,7-tetrahydrobenzofuran
Type Analytical Standard
Quantity 1 mL
Formula Weight 150.22
Percent Purity ≥99% (sum of enantiomers, GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.