accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17966-67-5

IUPAC Name: 4-methyl-2-(phenylformamido)pentanoic acid
Synonyms: 4-methyl-2-(phenylformamido)pentanoic acid
Molecular Weight (g/mol): 235.28
Molecular Formula: C13H17NO3
InChi Key: POLGZPYHEPOBFG-UHFFFAOYNA-N
SMILES: CC(C)CC(NC(=O)C1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results