Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzoyl-DL-leucine 99.0+%, TCI America™
SDP

Catalog No. B020625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B020625G 25 g
B020610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B020625G Supplier TCI America Supplier No. B020625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17966-67-5
Molecular Formula C13H17NO3
Molecular Weight (g/mol) 235.28
MDL Number MFCD00038295
InChI Key POLGZPYHEPOBFG-UHFFFAOYNA-N
PubChem CID 294887
IUPAC Name 4-methyl-2-(phenylformamido)pentanoic acid
SMILES CC(C)CC(NC(=O)C1=CC=CC=C1)C(O)=O

Specifications

Melting Point 141°C
Color White-Yellow
Quantity 25 g
Formula Weight 235.28
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzoyl-DL-leucine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.