accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOONNHOOCH3

CAS RN 179688-29-0

IUPAC Name: 6,7-bis(2-methoxyethoxy)-1,4-dihydroquinazolin-4-one
Synonyms: 6,7-bis(2-methoxyethoxy)-1H-quinazolin-4-one
Molecular Weight (g/mol): 294.31
Molecular Formula: C14H18N2O5
InChi Key: PMQWTUWLIGJTQD-UHFFFAOYSA-N
SMILES: COCCOC1=C(OCCOC)C=C2C(=O)N=CNC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results