Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6,7-Bis(2-methoxyethoxy)-3H-quinazolin-4-one 98.0+%, TCI America™
SDP

Catalog No. B42705G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42705G 5 g
B42701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B42705G Supplier TCI America Supplier No. B42705G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 179688-29-0
Molecular Formula C14H18N2O5
Molecular Weight (g/mol) 294.307
MDL Number MFCD02678087
InChI Key PMQWTUWLIGJTQD-UHFFFAOYSA-N
Synonym 6,7-Bis(2-methoxyethoxy)-4(3H)-quinazolinone, 6,7-Bis(2-methoxyethoxy)-3,4-dihydroquinazolin-4-one, 6,7-Bis(2-methoxyethoxy)-4-hydroxyquinazoline, 6,7-Bis(2-methoxyethoxy)-4-quinazolinol
PubChem CID 2809007
IUPAC Name 6,7-bis(2-methoxyethoxy)-1H-quinazolin-4-one
SMILES COCCOC1=C(C=C2C(=C1)C(=O)N=CN2)OCCOC

Specifications

Melting Point 192°C
Quantity 5 g
Formula Weight 294.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6,7-Bis(2-methoxyethoxy)-3H-quinazolin-4-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.