accessibility menu, dialog, popup

UserName

Viewing profile as user:
BH3CCH3H3CHNa

CAS RN 17979-81-6

IUPAC Name: sodium triethylboranuide
Synonyms: sodium triethylborohydride
Molecular Weight (g/mol): 121.99
Molecular Formula: C6H16BNa
InChi Key: YDTZLEUIYNMRLQ-UHFFFAOYSA-N
SMILES: [Na+].CC[BH-](CC)CC
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results