accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 17989-90-1

CH2OOH2COOBa

CAS RN 17989-90-1

IUPAC Name: barium(2+) bis(prop-2-enoate)
Synonyms: barium(2+) diacrylate
Molecular Weight (g/mol): 279.44
Molecular Formula: C6H6BaO4
InChi Key: ONQPQIFFWHMGGE-UHFFFAOYSA-L
SMILES: [Ba++].[O-]C(=O)C=C.[O-]C(=O)C=C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results