accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 179899-07-1

IUPAC Name: (4-fluoro-2-methoxyphenyl)boronic acid
Synonyms: 4-fluoro-2-methoxyphenylboronic acid
Molecular Weight (g/mol): 169.95
Molecular Formula: C7H8BFO3
InChi Key: ADJBXDCXYMCCAD-UHFFFAOYSA-N
SMILES: COC1=CC(F)=CC=C1B(O)O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results