Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluoro-2-methoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. F10625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F1062-5G 5 g
F10621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F1062-5G Supplier TCI America Supplier No. F10625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 179899-07-1
Molecular Formula C7H8BFO3
Molecular Weight (g/mol) 169.95
MDL Number MFCD03788423
InChI Key ADJBXDCXYMCCAD-UHFFFAOYSA-N
PubChem CID 2782672
IUPAC Name (4-fluoro-2-methoxyphenyl)boronic acid
SMILES COC1=CC(F)=CC=C1B(O)O

Specifications

Melting Point 136°C
Color White
Quantity 5 g
Synonym 4-fluoro-2-methoxyphenyl boronic acid, 2-methoxy-4-fluorophenylboronic acid, 4-fluoro-2-methoxybenzeneboronic acid, 4-fluoro-2-methoxy-phenyl boronic acid, 4-fluoro-2-methoxyphenyl-boronic acid
Formula Weight 169.95
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.