accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH2OOFFFFFFFFF

CAS RN 1799-84-4

IUPAC Name: 3,3,4,4,5,5,6,6,6-nonafluorohexyl 2-methylprop-2-enoate
Synonyms: 3,3,4,4,5,5,6,6,6-nonafluorohexyl 2-methylprop-2-enoate
Molecular Weight (g/mol): 332.17
Molecular Formula: C10H9F9O2
InChi Key: TYNRPOFACABVSI-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results