Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1H,1H,2H,2H-Nonafluorohexyl Methacrylate (stabilized with MEHQ) 98.0+%, TCI America™
SDP

Catalog No. N101425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N101425G 25 g
N10145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N101425G Supplier TCI America Supplier No. N101425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1799-84-4
Molecular Formula C10H9F9O2
Molecular Weight (g/mol) 332.17
MDL Number MFCD00236094
InChI Key TYNRPOFACABVSI-UHFFFAOYSA-N
Synonym Methacrylic Acid 1H,1H,2H,2H-Nonafluorohexyl Ester, 3,3,4,4,5,5,6,6,6-Nonafluorohexyl Methacrylate, Methacrylic Acid 3,3,4,4,5,5,6,6,6-Nonafluorohexyl Ester
PubChem CID 74534
IUPAC Name 3,3,4,4,5,5,6,6,6-nonafluorohexyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Color Colorless
Boiling Point 62°C
Quantity 25 g
Formula Weight 332.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1H,1H,2H,2H-Nonafluorohexyl Methacrylate (stabilized with MEHQ)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.