accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COSiCH2OH3COCH3

CAS RN 18001-13-3

IUPAC Name: (4-ethenylphenyl)trimethoxysilane
Synonyms: (4-ethenylphenyl)trimethoxysilane
Molecular Weight (g/mol): 224.33
Molecular Formula: C11H16O3Si
InChi Key: LTQBNYCMVZQRSD-UHFFFAOYSA-N
SMILES: CO[Si](OC)(OC)C1=CC=C(C=C)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results