Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethoxy(4-vinylphenyl)silane 97.0+%, TCI America™
SDP

Catalog No. T31855G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31855G 5 g
T318525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T31855G Supplier TCI America Supplier No. T31855G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18001-13-3
Molecular Formula C11H16O3Si
Molecular Weight (g/mol) 224.33
MDL Number MFCD21596662
InChI Key LTQBNYCMVZQRSD-UHFFFAOYSA-N
PubChem CID 15606295
IUPAC Name (4-ethenylphenyl)trimethoxysilane
SMILES CO[Si](OC)(OC)C1=CC=C(C=C)C=C1

Specifications

Color Yellow
Boiling Point 77°C
Quantity 5 g
Formula Weight 224.33
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethoxy(4-vinylphenyl)silane
EINECSNumber (3)-4371
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.