accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 180161-14-2

IUPAC Name: 6-fluoro-3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-indole
Synonyms: 6-fluoro-3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-indole
Molecular Weight (g/mol): 216.26
Molecular Formula: C13H13FN2
InChi Key: FBWKDFSEFVFKFY-UHFFFAOYSA-N
SMILES: FC1=CC=C2C(NC=C2C2=CCNCC2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results