Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Fluoro-3-(1,2,3,6-tetrahydro-4-pyridyl)indole, 97%, Thermo Scientific Chemicals

Catalog No. AAH6392006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6392006 5 g
AAH6392003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6392006 Supplier Thermo Scientific Chemicals Supplier No. H6392006

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 180161-14-2
Molecular Formula C13H13FN2
Molecular Weight (g/mol) 216.26
MDL Number MFCD04114772
InChI Key FBWKDFSEFVFKFY-UHFFFAOYSA-N
Synonym 6-fluoro-3-1,2,3,6-tetrahydropyridin-4-yl-1h-indole, 6-fluoro-3-1,2,3,6-tetrahydro-pyridin-4-yl-1h-indole, 6-fluoro-3-1,2,3,6-tetrahydro-4-pyridinyl-1h-indole, 6-fluoro-3-4-1,2,5,6-tetrahydropyridyl indole, 4-6-fluoroindol-3-yl-1,2,5,6-tetrahydropyridine, 6-fluoro-3-1,2,3,6-tetrahydro4-pyridinyl-1h-indole, 3-1,2,3,6-tetrahydropyridine-4-yl-6-fluoro-1h-indole, 4-6-fluoro-1h-indol-3-yl-1,2,3,6-tetrahydropyridine
PubChem CID 11053072
IUPAC Name 6-fluoro-3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-indole
SMILES FC1=CC=C2C(NC=C2C2=CCNCC2)=C1

Specifications

Color Yellow to Brown
Quantity 5 g
Beilstein 8627107
Formula Weight 216.25
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 6-Fluoro-3-(1,2,3,6-tetrahydro-4-pyridyl)indole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.