CAS RN 18035-33-1
IUPAC Name:
trichloro(6-phenylhexyl)silane
Synonyms:
trichloro(6-phenylhexyl)silane
Molecular Weight (g/mol):
295.7
Molecular Formula:
C12H17Cl3Si
InChi Key:
ODIAUNNUTSSHMI-UHFFFAOYSA-N
SMILES:
Cl[Si](Cl)(Cl)CCCCCCC1=CC=CC=C1
1
–
1
of
1
results
1
Trichloro(6-phenylhexyl)silane 98.0+%, TCI America™
CAS: 18035-33-1 Molecular Formula: C12H17Cl3Si Molecular Weight (g/mol): 295.70 MDL Number: MFCD09953885 InChI Key: ODIAUNNUTSSHMI-UHFFFAOYSA-N Synonym: (6-Phenylhexyl)trichlorosilane PubChem CID: 53427322 IUPAC Name: trichloro(6-phenylhexyl)silane SMILES: Cl[Si](Cl)(Cl)CCCCCCC1=CC=CC=C1