Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloro(6-phenylhexyl)silane 98.0+%, TCI America™
SDP

Catalog No. T28545G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28545G 5 g
T285425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T28545G Supplier TCI America Supplier No. T28545G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18035-33-1
Molecular Formula C12H17Cl3Si
Molecular Weight (g/mol) 295.70
MDL Number MFCD09953885
InChI Key ODIAUNNUTSSHMI-UHFFFAOYSA-N
Synonym (6-Phenylhexyl)trichlorosilane
PubChem CID 53427322
IUPAC Name trichloro(6-phenylhexyl)silane
SMILES Cl[Si](Cl)(Cl)CCCCCCC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 92°C
Quantity 5 g
UN Number 2987
Formula Weight 295.70
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Trichloro(6-phenylhexyl)silane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.