accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18063-03-1

IUPAC Name: 2,6-difluorobenzamide
Synonyms: 2,6-difluorobenzamide
Molecular Weight (g/mol): 157.12
Molecular Formula: C7H5F2NO
InChi Key: AVRQBXVUUXHRMY-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(F)C=CC=C1F
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results