Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Difluorobenzamide 97.0+%, TCI America™
SDP

Catalog No. D190725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D190725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D190725G Supplier TCI America Supplier No. D190725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18063-03-1
Molecular Formula C7H5F2NO
Molecular Weight (g/mol) 157.12
MDL Number MFCD00007972
InChI Key AVRQBXVUUXHRMY-UHFFFAOYSA-N
Synonym benzamide, 2,6-difluoro, 2,6-dfam, unii-sj2rn214mk, sj2rn214mk, 2,6-difluorobenzoylamide, 2,6 difluorobenzamide, 2.6-difluorobenzamide, pubchem10766, 2, 6 difluorobenzamide, 2,6 difluorobenzamide;
PubChem CID 87439
IUPAC Name 2,6-difluorobenzamide
SMILES C1=CC(=C(C(=C1)F)C(=O)N)F

Specifications

Melting Point 145°C
Color White
Quantity 25 g
Formula Weight 157.12
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Difluorobenzamide
EINECSNumber (3)-3776
RTECSNumber CV4355050
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.