accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18065-11-7

IUPAC Name: 1-(4-methoxybenzoyl)-2-methylpiperidine
Synonyms: 1-(4-methoxybenzoyl)-2-methylpiperidine
Molecular Weight (g/mol): 233.31
Molecular Formula: C14H19NO2
InChi Key: SGLSSNVBXPORFL-UHFFFAOYNA-N
SMILES: COC1=CC=C(C=C1)C(=O)N1CCCCC1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results