accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNNOH

CAS RN 18073-84-2

IUPAC Name: 2-(4-{3-[1-(2-hydroxyethyl)piperidin-4-yl]propyl}piperidin-1-yl)ethan-1-ol
Synonyms: 2-(4-{3-[1-(2-hydroxyethyl)piperidin-4-yl]propyl}piperidin-1-yl)ethanol
Molecular Weight (g/mol): 298.47
Molecular Formula: C17H34N2O2
InChi Key: VSSGEWPIFHKREK-UHFFFAOYSA-N
SMILES: OCCN1CCC(CCCC2CCN(CCO)CC2)CC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results