Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Bis[1-(2-hydroxyethyl)-4-piperidyl]propane 98.0+%, TCI America™
SDP

Catalog No. B085125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B085125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B085125G Supplier TCI America Supplier No. B085125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18073-84-2
Molecular Formula C17H34N2O2
Molecular Weight (g/mol) 298.471
MDL Number MFCD00023767
InChI Key VSSGEWPIFHKREK-UHFFFAOYSA-N
PubChem CID 87445
IUPAC Name 2-[4-[3-[1-(2-hydroxyethyl)piperidin-4-yl]propyl]piperidin-1-yl]ethanol
SMILES C1CN(CCC1CCCC2CCN(CC2)CCO)CCO

Specifications

Melting Point 95°C
Color White-Yellow
Boiling Point 219°C
Quantity 25 g
Formula Weight 298.47
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis[1-(2-hydroxyethyl)-4-piperidyl]propane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.