accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOOOOH3C(?)

CAS RN 1807549-00-3

IUPAC Name: ethyl 2-{6-[(4-methyl-5-oxo-2,5-dihydrofuran-2-yl)oxy]-3-oxo-3H-xanthen-9-yl}benzoate
Synonyms: ethyl 2-{3-[(4-methyl-5-oxo-2H-furan-2-yl)oxy]-6-oxoxanthen-9-yl}benzoate
Molecular Weight (g/mol): 456.45
Molecular Formula: C27H20O7
InChi Key: CHRFZINIRYDHJV-UHFFFAOYNA-N
SMILES: CCOC(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC(OC3OC(=O)C(C)=C3)=CC=C12
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)
pH 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)