Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Yoshimulactone Green (YLG) 98.0+%, TCI America™
SDP

Catalog No. E12381MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E12381MG 1 mg
E123810MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E12381MG Supplier TCI America Supplier No. E12381MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1807549-00-3
Molecular Formula C27H20O7
Molecular Weight (g/mol) 456.45
MDL Number MFCD29472531
InChI Key CHRFZINIRYDHJV-UHFFFAOYNA-N
Synonym 2-[6-[(2,5-Dihydro-4-methyl-5-oxo-2-furanyl)oxy]-3-oxo-3H-xanthen-9-yl]benzoic Acid Ethyl Ester, Ethyl 2-[6-[(2,5-Dihydro-4-methyl-5-oxo-2-furanyl)oxy]-3-oxo-3H-xanthen-9-yl]benzoate
PubChem CID 131637025
IUPAC Name ethyl 2-{6-[(4-methyl-5-oxo-2,5-dihydrofuran-2-yl)oxy]-3-oxo-3H-xanthen-9-yl}benzoate
SMILES CCOC(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC(OC3OC(=O)C(C)=C3)=CC=C12

Specifications

Melting Point 190°C
pH White to Pale Reddish Gray
Quantity 1 mg
Physical Form Crystal/Powder
Formula Weight 456.45
Percent Purity ≥98.0% (HPLC)
Chemical Name or Material Yoshimulactone Green (YLG)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.