CAS RN 18080-67-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Peroxiredoxin Inhibitor, Conoidin A
A cell-permeable nematicidal compound that covalently binds to the peroxidatic cysteine of peroxiredoxin II in T. gondii and irreversibly blocks its hyperperoxidation activity (IC50 = 23 μM).