Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Peroxiredoxin Inhibitor, Conoidin A

Catalog No. 5307460001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-074-60001 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-074-60001 Supplier MilliporeSigma™ Supplier No. 5307460001
Add to Cart
Add to Cart

A cell-permeable nematicidal compound that covalently binds to the peroxidatic cysteine of peroxiredoxin II in T. gondii and irreversibly blocks its hyperperoxidation activity (IC50 = 23 μM).

  • Packaged Under Inert Gas
  • Cell Permeable
  • Deactivate with 5% Bleach
TRUSTED_SUSTAINABILITY

Specifications

Color Yellowish Brown
Description 95% by NMR
Target Proteins Peroxiredoxin
CAS 18080-67-6
Molecular Formula C10H8Br2N2O2
Solubility DMSO
Content And Storage Protect from Light, Protect from Moisture
Form Solid
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.