CAS RN 1810-72-6
IUPAC Name:
2,6-dichloroquinoline
Synonyms:
2,6-dichloroquinoline
Molecular Weight (g/mol):
198.05
Molecular Formula:
C9H5Cl2N
InChi Key:
LPDFGLZUUCLXGM-UHFFFAOYSA-N
SMILES:
ClC1=CC=C2N=C(Cl)C=CC2=C1
1
–
1
of
1
results
1
2,6-Dichloroquinoline 98.0+%, TCI America™
CAS: 1810-72-6 Molecular Formula: C9H5Cl2N Molecular Weight (g/mol): 198.05 MDL Number: MFCD03427194 InChI Key: LPDFGLZUUCLXGM-UHFFFAOYSA-N PubChem CID: 607502 IUPAC Name: 2,6-dichloroquinoline SMILES: ClC1=CC=C2N=C(Cl)C=CC2=C1