accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18107-18-1

IUPAC Name: (diazomethyl)trimethylsilane
Synonyms: trimethylsilyldiazomethane
Molecular Weight (g/mol): 114.22
Molecular Formula: C4H10N2Si
InChi Key: ONDSBJMLAHVLMI-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C=[N+]=[N-]
Molecular Weight (g/mol) 
  • (4)
  • (3)
Physical Form 
  • (3)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (4)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (4)
  • (3)
  • (3)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)
  • (4)