CAS RN 18113-04-7
IUPAC Name:
2-chloro-5-methoxyphenol
Synonyms:
2-chloro-5-methoxyphenol
Molecular Weight (g/mol):
158.58
Molecular Formula:
C7H7ClO2
InChi Key:
KULSVCANYQIOFD-UHFFFAOYSA-N
SMILES:
COC1=CC=C(Cl)C(O)=C1
1
–
1
of
1
results
1
2-Chloro-5-methoxyphenol 97.0+%, TCI America™
CAS: 18113-04-7 Molecular Formula: C7H7ClO2 Molecular Weight (g/mol): 158.581 MDL Number: MFCD00086345 InChI Key: KULSVCANYQIOFD-UHFFFAOYSA-N Synonym: 4-Chloro-3-hydroxyanisole PubChem CID: 140336 IUPAC Name: 2-chloro-5-methoxyphenol SMILES: COC1=CC(=C(C=C1)Cl)O