Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-methoxyphenol 97.0+%, TCI America™
SDP

Catalog No. C272025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C272025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C272025G Supplier TCI America Supplier No. C272025G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18113-04-7
Molecular Formula C7H7ClO2
Molecular Weight (g/mol) 158.581
MDL Number MFCD00086345
InChI Key KULSVCANYQIOFD-UHFFFAOYSA-N
Synonym 4-Chloro-3-hydroxyanisole
PubChem CID 140336
IUPAC Name 2-chloro-5-methoxyphenol
SMILES COC1=CC(=C(C=C1)Cl)O

Specifications

Color Yellow
Boiling Point 125°C
Quantity 25 g
Formula Weight 158.58
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-5-methoxyphenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.