CAS RN 1811539-37-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ Janelia Fluor™ 525, Haloalkane
Cell-permeable fluorogenic fluorescent dye with a chloroalkane handle. Application: live-cell imaging as a self-labeling tag substrate