CAS RN 1813-60-1
IUPAC Name:
tetrabutylphosphanium; tetrafluoroboranuide
Synonyms:
tetrabutylphosphonium tetrafluoroborate
Molecular Weight (g/mol):
346.24
Molecular Formula:
C16H36BF4P
InChi Key:
BYCHYJSDSHOWIP-UHFFFAOYSA-N
SMILES:
F[B-](F)(F)F.CCCC[P+](CCCC)(CCCC)CCCC
1
–
1
of
1
results
1
Tetrabutylphosphonium Tetrafluoroborate 97.0+%, TCI America™
CAS: 1813-60-1 Molecular Formula: C16H36BF4P Molecular Weight (g/mol): 346.24 MDL Number: MFCD00270144 InChI Key: BYCHYJSDSHOWIP-UHFFFAOYSA-N PubChem CID: 13669093 IUPAC Name: tetrabutylphosphanium; tetrafluoroboranuide SMILES: F[B-](F)(F)F.CCCC[P+](CCCC)(CCCC)CCCC