Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrabutylphosphonium Tetrafluoroborate 97.0+%, TCI America™
SDP

Catalog No. T200625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T200625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T200625G Supplier TCI America Supplier No. T200625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1813-60-1
Molecular Formula C16H36BF4P
Molecular Weight (g/mol) 346.24
MDL Number MFCD00270144
InChI Key BYCHYJSDSHOWIP-UHFFFAOYSA-N
PubChem CID 13669093
IUPAC Name tetrabutylphosphanium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.CCCC[P+](CCCC)(CCCC)CCCC

Specifications

Melting Point 98°C
Color White
Quantity 25 g
UN Number 1759
Formula Weight 346.24
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylphosphonium Tetrafluoroborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.