accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 181311-44-4

FFFFFOOOOOONNH(S)CH3H3CCH3

CAS RN 181311-44-4

IUPAC Name: tert-butyl 3-[(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-oxo-3-(2,3,4,5,6-pentafluorophenoxy)propyl]-1H-indole-1-carboxylate
Synonyms: tert-butyl 3-[(2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-oxo-3-(2,3,4,5,6-pentafluorophenoxy)propyl]indole-1-carboxylate
Molecular Weight (g/mol): 692.64
Molecular Formula: C37H29F5N2O6
InChi Key: IDYZGIYCOLJXRE-SANMLTNESA-N
SMILES: CC(C)(C)OC(=O)N1C=C(C[C@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(=O)OC2=C(F)C(F)=C(F)C(F)=C2F)C2=CC=CC=C12
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)