accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18138-03-9

IUPAC Name: 2-propylpyrazine
Synonyms: 2-propylpyrazine
Molecular Weight (g/mol): 122.17
Molecular Formula: C7H10N2
InChi Key: DJLLTFRHLPVCEL-UHFFFAOYSA-N
SMILES: CCCC1=CN=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results