Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propylpyrazine 98.0+%, TCI America™
SDP

Catalog No. P18335G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P18335G Supplier TCI America Supplier No. P18335G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18138-03-9
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00049214
InChI Key DJLLTFRHLPVCEL-UHFFFAOYSA-N
PubChem CID 87466
IUPAC Name 2-propylpyrazine
SMILES CCCC1=CN=CC=N1

Specifications

Color Yellow
Boiling Point 174°C
Quantity 5 g
Formula Weight 122.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Propylpyrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.