accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18146-62-8

IUPAC Name: tributylsulfanium iodide
Synonyms: tributylsulfanium iodide
Molecular Weight (g/mol): 330.31
Molecular Formula: C12H27IS
InChi Key: HUCQSHDLMUWBPS-UHFFFAOYSA-M
SMILES: [I-].CCCC[S+](CCCC)CCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results