Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tributylsulfonium Iodide 96.0+%, TCI America™
SDP

Catalog No. T15641G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15641G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T15641G Supplier TCI America Supplier No. T15641G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18146-62-8
Molecular Formula C12H27IS
Molecular Weight (g/mol) 330.312
MDL Number MFCD00671571
InChI Key HUCQSHDLMUWBPS-UHFFFAOYSA-M
PubChem CID 21988031
IUPAC Name tributylsulfanium;iodide
SMILES CCCC[S+](CCCC)CCCC.[I-]

Specifications

Melting Point 93°C
Color Yellow
Quantity 1 g
Formula Weight 330.31
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tributylsulfonium Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.