CAS RN 18156-74-6
N-Trimethylsilylimidazole, 99%, MP Biomedicals™
CAS: 18156-74-6 Molecular Formula: C6H12N2Si Molecular Weight (g/mol): 140.26 MDL Number: MFCD00005280 InChI Key: YKFRUJSEPGHZFJ-UHFFFAOYSA-N Synonym: n-trimethylsilyl imidazole,1-trimethylsilyl imidazole,1-trimethylsilyl-1h-imidazole,tsim,1h-imidazole, 1-trimethylsilyl,n-trimethylsilylimidazole,trimethylsilyl imidazole,1-trimethylsilylimidazole,n-trimethylsilyl imidazol,imidazole, 1-trimethylsilyl PubChem CID: 28925 ChEBI: CHEBI:85063 IUPAC Name: 1-(trimethylsilyl)-1H-imidazole SMILES: C[Si](C)(C)N1C=CN=C1
1-(Trimethylsilyl)imidazole, 97%
CAS: 18156-74-6 Molecular Formula: C6H12N2Si Molecular Weight (g/mol): 140.26 MDL Number: MFCD00005280 InChI Key: YKFRUJSEPGHZFJ-UHFFFAOYSA-N Synonym: n-trimethylsilyl imidazole,1-trimethylsilyl imidazole,1-trimethylsilyl-1h-imidazole,tsim,1h-imidazole, 1-trimethylsilyl,n-trimethylsilylimidazole,trimethylsilyl imidazole,1-trimethylsilylimidazole,n-trimethylsilyl imidazol,imidazole, 1-trimethylsilyl PubChem CID: 28925 ChEBI: CHEBI:85063 IUPAC Name: imidazol-1-yl(trimethyl)silane SMILES: C[Si](C)(C)N1C=CN=C1
1-(Trimethylsilyl)imidazole, Approx. 97%, Spectrum™ Chemical
CAS: 18156-74-6 Molecular Formula: C6H12N2Si Molecular Weight (g/mol): 140.26 MDL Number: MFCD00005280 InChI Key: YKFRUJSEPGHZFJ-UHFFFAOYSA-N IUPAC Name: 1-(trimethylsilyl)-1H-imidazole SMILES: C[Si](C)(C)N1C=CN=C1
N-(Trimethylsilyl)imidazole, 97%
CAS: 18156-74-6 Molecular Formula: C6H12N2Si Molecular Weight (g/mol): 140.26 MDL Number: MFCD00005280 InChI Key: YKFRUJSEPGHZFJ-UHFFFAOYSA-N Synonym: n-trimethylsilyl imidazole,1-trimethylsilyl imidazole,1-trimethylsilyl-1h-imidazole,tsim,1h-imidazole, 1-trimethylsilyl,n-trimethylsilylimidazole,trimethylsilyl imidazole,1-trimethylsilylimidazole,n-trimethylsilyl imidazol,imidazole, 1-trimethylsilyl PubChem CID: 28925 ChEBI: CHEBI:85063 IUPAC Name: imidazol-1-yl(trimethyl)silane SMILES: C[Si](C)(C)N1C=CN=C1
N-Trimethylsilylimidazole 98.0+%, TCI America™
CAS: 18156-74-6 Molecular Formula: C6H12N2Si Molecular Weight (g/mol): 140.26 MDL Number: MFCD00005280 InChI Key: YKFRUJSEPGHZFJ-UHFFFAOYSA-N Synonym: n-trimethylsilyl imidazole,1-trimethylsilyl imidazole,1-trimethylsilyl-1h-imidazole,tsim,1h-imidazole, 1-trimethylsilyl,n-trimethylsilylimidazole,trimethylsilyl imidazole,1-trimethylsilylimidazole,n-trimethylsilyl imidazol,imidazole, 1-trimethylsilyl PubChem CID: 28925 ChEBI: CHEBI:85063 IUPAC Name: 1-(trimethylsilyl)-1H-imidazole SMILES: C[Si](C)(C)N1C=CN=C1
TMS-Imidazole (=N-Trimethylsilylimidazole) 98.0+%, TCI America™
CAS: 18156-74-6 Molecular Formula: C6H12N2Si Molecular Weight (g/mol): 140.26 MDL Number: MFCD00005280 InChI Key: YKFRUJSEPGHZFJ-UHFFFAOYSA-N Synonym: n-trimethylsilyl imidazole,1-trimethylsilyl imidazole,1-trimethylsilyl-1h-imidazole,tsim,1h-imidazole, 1-trimethylsilyl,n-trimethylsilylimidazole,trimethylsilyl imidazole,1-trimethylsilylimidazole,n-trimethylsilyl imidazol,imidazole, 1-trimethylsilyl PubChem CID: 28925 ChEBI: CHEBI:85063 IUPAC Name: 1-(trimethylsilyl)-1H-imidazole SMILES: C[Si](C)(C)N1C=CN=C1
Sigma Aldrich 1-(Trimethylsilyl)imidazole
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich 1-(Trimethylsilyl)imidazole
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.