Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-(Trimethylsilyl)imidazole

Catalog No. 1535835ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-358-35ML 5 mL
15-358-325ML 25 mL
15-358-3100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 15-358-35ML Supplier Sigma Aldrich Supplier No. 1535835ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Silylating reagent for the protection of hydroxyl groups in the presence of amine functionalities.

Specifications

CAS 18156-74-6
Density 0.956 g/mL (at 25°C (literature))
Boiling Point 93°C to 94°C (14 mmHg)
Molecular Formula C6H12N2Si
Refractive Index n20/D 1.475 (literature)
Linear Formula C6H12N2Si
MDL Number MFCD00005280
Quantity 5 mL
Synonym N-Trimethylsilylimidazole; TMSI; TSIM
Molecular Weight (g/mol) 140.26
Percent Purity 96%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.