accessibility menu, dialog, popup

UserName

Viewing profile as user:
NClFFFFF

CAS RN 1816283-20-1

IUPAC Name: 4-chloro-2-(1,1,2,2,2-pentafluoroethyl)pyridine
Synonyms: 4-chloro-2-(1,1,2,2,2-pentafluoroethyl)pyridine
Molecular Weight (g/mol): 231.55
Molecular Formula: C7H3ClF5N
InChi Key: ASGAWEVNGFJCIS-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C1=NC=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results