accessibility menu, dialog, popup

UserName

Viewing profile as user:
NFFFFFCl

CAS RN 1816286-90-4

IUPAC Name: 3-chloro-4-(1,1,2,2,2-pentafluoroethyl)pyridine
Synonyms: 3-chloro-4-(1,1,2,2,2-pentafluoroethyl)pyridine
Molecular Weight (g/mol): 231.55
Molecular Formula: C7H3ClF5N
InChi Key: RMNYIIRMELWLLC-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C1=C(Cl)C=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results