accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONOHNNFFFF

CAS RN 181629-93-6

IUPAC Name: N-[3-fluoro-5-(pyridin-3-yl)phenyl]-5-methoxy-6-(trifluoromethyl)-2,3-dihydro-1H-indole-1-carboxamide
Synonyms: N-[3-fluoro-5-(pyridin-3-yl)phenyl]-5-methoxy-6-(trifluoromethyl)-2,3-dihydroindole-1-carboxamide
Molecular Weight (g/mol): 431.39
Molecular Formula: C22H17F4N3O2
InChi Key: RRJLJKRFFRZRAF-UHFFFAOYSA-N
SMILES: COC1=C(C=C2N(CCC2=C1)C(=O)NC1=CC(F)=CC(=C1)C1=CN=CC=C1)C(F)(F)F
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results