accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18166-37-5

IUPAC Name: triethoxy(hexyl)silane
Synonyms: triethoxy(hexyl)silane
Molecular Weight (g/mol): 248.44
Molecular Formula: C12H28O3Si
InChi Key: WUMSTCDLAYQDNO-UHFFFAOYSA-N
SMILES: CCCCCC[Si](OCC)(OCC)OCC
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results