Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexyltriethoxysilane, TCI America™
SDP

Catalog No. H115825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H115825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H115825G Supplier TCI America Supplier No. H115825G

Chemical Identifiers

CAS 18166-37-5
Molecular Formula C12H28O3Si
Molecular Weight (g/mol) 248.44
MDL Number MFCD00087744
InChI Key WUMSTCDLAYQDNO-UHFFFAOYSA-N
Synonym hexyltriethoxysilane, triethoxy hexyl silane, hexyl triethoxysilane, n-hexyltriethoxysilane, hexyl-triethoxysilane, acmc-209ei3
PubChem CID 4437318
IUPAC Name triethoxy(hexyl)silane
SMILES CCCCCC[Si](OCC)(OCC)OCC

Specifications

Color Colorless
Boiling Point 115°C
Quantity 25 g
Formula Weight 248.44
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexyltriethoxysilane
EINECSNumber (2)-2052
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.