accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1817-73-8

IUPAC Name: 2-bromo-4,6-dinitroaniline
Synonyms: 2-bromo-4,6-dinitroaniline
Molecular Weight (g/mol): 262.02
Molecular Formula: C6H4BrN3O4
InChi Key: KWMDHCLJYMVBNS-UHFFFAOYSA-N
SMILES: NC1=C(Br)C=C(C=C1[N+]([O-])=O)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)