Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4,6-dinitroaniline, 97%, Thermo Scientific™

Catalog No. AAB2290522
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2290522 100 g
AAB2290514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2290522 Supplier Thermo Scientific Chemicals Supplier No. B2290522

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1817-73-8
Molecular Formula C6H4BrN3O4
Molecular Weight (g/mol) 262.019
MDL Number MFCD00007146
InChI Key KWMDHCLJYMVBNS-UHFFFAOYSA-N
Synonym 6-bromo-2,4-dinitroaniline, bromo dna, 2,4-dinitro-6-bromoaniline, benzenamine, 2-bromo-4,6-dinitro, 2-bromo-4,6-dinitroaminobenzene, 2,4-dinitro-6-bromanilin, aniline, 2,4-dinitro-6-bromo, aniline, 2-bromo-4,6-dinitro, ccris 820, 2,4-dinitro-6-bromanilin czech
PubChem CID 15752
IUPAC Name 2-bromo-4,6-dinitroaniline
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])N)Br)[N+](=O)[O-]

Specifications

Melting Point 151°C to 153°C
Quantity 100 g
Formula Weight 262.02
Percent Purity 97%
Chemical Name or Material 2-Bromo-4,6-dinitroaniline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H341
Suspected of causing genetic defects.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-329-2
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.