accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONO

CAS RN 1817-77-2

IUPAC Name: 1-benzyl-4-nitrobenzene
Synonyms: 1-benzyl-4-nitrobenzene
Molecular Weight (g/mol): 213.24
Molecular Formula: C13H11NO2
InChi Key: IDSGFSCSMXRJON-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(CC2=CC=CC=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results