Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrodiphenylmethane 98.0+%, TCI America™
SDP

Catalog No. N056425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N056425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N056425G Supplier TCI America Supplier No. N056425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1817-77-2
Molecular Formula C13H11NO2
Molecular Weight (g/mol) 213.236
MDL Number MFCD00024783
InChI Key IDSGFSCSMXRJON-UHFFFAOYSA-N
PubChem CID 137225
IUPAC Name 1-benzyl-4-nitrobenzene
SMILES C1=CC=C(C=C1)CC2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Color Yellow
Boiling Point 160°C
Quantity 25 g
Formula Weight 213.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Nitrodiphenylmethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.